missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZD3965, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
AZD3965 is a selective MCT1 inhibitor with a Ki of 1.6 nM, showing 6-fold selectivity over MCT2.
Specifications
Specifications
| Chemical Name or Material | AZD3965 |
| Physical Form | Solid |
| CAS | 1448671-31-5 |
| Color | White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H24F3N5O5S |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : 100 mg/mL (193.98 mM; Need ultrasonic) |
| SMILES | O=C1N(C(C)C)C2=C(C(C(N3OC[C@@](C)(O)C3)=O)=C(CC4=C(C)NN=C4C(F)(F)F)S2)C(N1C)=O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction