missing translation for 'onlineSavingsMsg'
Learn More

AZD3839 free base, MedChemExpress

Product Code. 16481847
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16481847 50 mg 50mg
16468377 5 mg 5mg
16463356 25 mg 25mg
16425688 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481847 Supplier Medchem Express Supplier No. HY13438

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD3839 free base is a potent and selective orally active, brain-permeable BACE1 inhibitor (Ki=26 nM).

AZD3839 free base shows 14 and >1000-fold selectivity against BACE2 and cathepsin D, respectively. AZD3839 free base exhibits dose- and time-dependent lowering of plasma, brain, and cerebrospinal fluid Aβ levels in mouse, guinea pig, and non-human primate. AZD3839 free base can be used for the research of Alzheimer's disease.

Specifications

Chemical Name or Material AZD3839 free base
CAS 1227163-84-9
Color White
Purity Grade Notes Research
Molecular Formula C24H16F3N5
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (289.75 mM; Need ultrasonic)
SMILES FC1=C2C([C@](C3=CC=CC(C4=CN=CN=C4)=C3)(C5=CC(C(F)F)=NC=C5)N=C2N)=CC=C1
Molecular Weight (g/mol) 431.41
Formula Weight 431.41
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.