missing translation for 'onlineSavingsMsg'
Learn More

AZD2932, MedChemExpress

Product Code. 16467478
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16467478 10 mg 10mg
16459957 5 mg 5mg
16427446 100 mg 100mg
16413507 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16467478 Supplier Medchem Express Supplier No. HY18179

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD2932 is a potent and multi-targeted kinase inhibitor VEGFR2, PDGFβ, Flt-3 and c-Kit with IC50s of 8, 4, 7 and 9 nM in cell assay, respectively.

AZD2932 is a potent and multi-targeted kinase inhibitor VEGFR2, PDGFβ, Flt-3 and c-Kit with IC50s of 8, 4, 7 and 9 nM in cell assay, respectively.

Specifications

Chemical Name or Material AZD2932
CAS 883986-34-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H25N5O4
Quantity 10 mg
Solubility Information DMSO : 10 mg/mL (22.35 mM; Need ultrasonic)
SMILES O=C(NC1=CN(C(C)C)N=C1)CC2=CC=C(OC3=C4C=C(OC)C(OC)=CC4=NC=N3)C=C2
Molecular Weight (g/mol) 447.49
Formula Weight 447.49
Percent Purity 96.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.