missing translation for 'onlineSavingsMsg'
Learn More

AZD2098, MedChemExpress

Product Code. 16460139
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460139 10 mg 10mg
16412338 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16460139 Supplier Medchem Express Supplier No. HYU00064
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AZD2098 is a potent and selective CC-chemokine receptor 4 (CCR4) inhibitor with pIC50s of 7.8, 8.0, 8.0 and 7.6 for human, rat, mouse and dog respectively, used for asthma research.

AZD2098 is a potent and selective CC-chemokine receptor 4 (CCR4) inhibitor with pIC50s of 7.8, 8.0, 8.0 and 7.6 for human, rat, mouse and dog respectively, used for asthma research.

Specifications

Chemical Name or Material AZD2098
CAS 566203-88-1
Color White
Purity Grade Notes Research
Molecular Formula C11H9Cl2N3O3S
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (299.24 mM; Need ultrasonic)
SMILES O=S(C1=CC=CC(Cl)=C1Cl)(NC2=NC=CN=C2OC)=O
Molecular Weight (g/mol) 334.18
Formula Weight 334.18
Percent Purity 99.86%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.