missing translation for 'onlineSavingsMsg'
Learn More

AZD1981, MedChemExpress

Product Code. 16489467
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16489467 5 mg 5mg
16457006 100 mg 100mg
16436928 10 mg 10mg
16422987 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16489467 Supplier Medchem Express Supplier No. HY15950
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AZD1981 is a potent and selective CRTh2 antagonist; displaces radio-labelled PGD2 from human recombinant DP2 with high potency (pIC50 = 8.4).

AZD1981 is a potent and selective CRTh2 antagonist

Specifications

Chemical Name or Material AZD1981
CAS 802904-66-1
Color Gray
Purity Grade Notes Research
Molecular Formula C19H17ClN2O3S
Quantity 5 mg
Solubility Information DMSO : ≥ 31 mg/mL (79.72 mM)
SMILES O=C(O)CN1C(C)=C(SC2=CC=C(Cl)C=C2)C3=C1C=CC=C3NC(C)=O
Molecular Weight (g/mol) 388.87
Formula Weight 388.87
Percent Purity 99.81%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.