missing translation for 'onlineSavingsMsg'
Learn More

AZD1283, MedChemExpress

Product Code. 16456946
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16456946

Brand: Medchem Express HY15799

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD1283 is a potent antagonist of the P2Y12 receptor with EC50 of 3.0 ug/kg/min, TI >10; with binding IC50 of 11 nM.

AZD1283 is a potent antagonist of the P2Y12 receptor with EC50 of 3.0 ug/kg/min, TI >10

Specifications

Chemical Name or Material AZD1283
CAS 919351-41-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C23H26N4O5S
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (212.52 mM; Need ultrasonic)
SMILES CC1=C(C(OCC)=O)C=C(C#N)C(N2CCC(C(NS(=O)(CC3=CC=CC=C3)=O)=O)CC2)=N1
Molecular Weight (g/mol) 470.54
Formula Weight 470.54
Percent Purity 98.01%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.