missing translation for 'onlineSavingsMsg'
Learn More

AZD1080, MedChemExpress

Product Code. 16485888
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16485888 10 mg 10mg
16482037 50 mg 50mg
16476346 100 mg 100mg
16408537 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16485888 Supplier Medchem Express Supplier No. HY13862

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD1080 is a potent and selective GSK3 inhibitor. AZD1080 inhibits recombinant human GSK3α and GSK3β with pKi (IC50) of 8.2 (6.9 nM) and 7.5 (31 nM), respectively.

AZD1080 is a potent and selective GSK3 inhibitor. AZD1080 inhibits recombinant human GSK3α and GSK3β with pKi (IC50) of 8.2 (6.9 nM) and 7.5 (31 nM), respectively.

Specifications

Chemical Name or Material AZD1080
Physical Form Solid
CAS 612487-72-6
Color Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C19H18N4O2
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information DMSO : 21.35 mg/mL (63.85 mM; Need ultrasonic and warming)
SMILES OC(N1)=C(C2=CC=C(CN3CCOCC3)C=N2)C4=C1C=CC(C#N)=C4
Molecular Weight (g/mol) 334.37
Formula Weight 334.37
Percent Purity 96.58%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.