missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZD0156, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
AZD0156 is a potent, selective and orally active ATM inhibitor with an IC50 of 0.58 nM. AZD0156 inhibits the ATM-mediated signaling, prevents DNA damage checkpoint activation, disrupts DNA damage repair, and induces tumor cell apoptosis.
Specifications
Specifications
| Chemical Name or Material | AZD0156 |
| CAS | 1821428-35-6 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C26H31N5O3 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 20 mg/mL (43.33 mM; ultrasonic and adjust pH to 3 with HCl) |
| SMILES | O=C(N1C2CCOCC2)N(C)C3=C1C4=CC(C5=CC=C(OCCCN(C)C)N=C5)=CC=C4N=C3 |
| Molecular Weight (g/mol) | 461.56 |
| Formula Weight | 461.56 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction