missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZD-8055, MedChemExpress
Description
AZD-8055 is a potent, selective, and orally bioavailable ATP-competitive mTOR kinase inhibitor with an IC50 of 0.8 nM. AZD-8055 inhibits both mTORC1 and mTORC2.
Specifications
Specifications
| Chemical Name or Material | AZD-8055 |
| CAS | 1009298-09-2 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C25H31N5O4 |
| Quantity | 50 mg |
| Solubility Information | DMSO : 33.33 mg/mL (71.59 mM; Need ultrasonic) |
| SMILES | OCC1=CC(C2=NC3=NC(N4CCOC[C@@H]4C)=NC(N5[C@@H](C)COCC5)=C3C=C2)=CC=C1OC |
| Molecular Weight (g/mol) | 465.54 |
| Formula Weight | 465.54 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?