missing translation for 'onlineSavingsMsg'
Learn More

AZD-6280, MedChemExpress

Product Code. 16421889
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16421889 1 mg 1mg
16490408 5 mg 5mg
16457948 10 mg 10mg
16400616 20 mg 20mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16421889 Supplier Medchem Express Supplier No. HY19872
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AZD-6280 is a selective GABAA(α2/3) receptor modulator, used for treatment of generalized anxiety disorder.

AZD-6280 is a selective GABAA(α2/3) receptor modulator, used for treatment of generalized anxiety disorder.

Specifications

Chemical Name or Material AZD-6280
CAS 942436-93-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C20H22N4O3
Quantity 1 mg
Solubility Information DMSO : 62.5 mg/mL (170.57 mM; Need ultrasonic)
SMILES O=C(C1=NN=C2C(C3=CC(OC)=CC=C3OC)=CC=CC2=C1N)NCCC
Molecular Weight (g/mol) 366.41
Formula Weight 366.41
Percent Purity 98.17%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.