missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZD 3988

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AZD 3988 is a potent and selective diacylglycerol acyltransferase (DGAT-1) inhibitor (IC50 = 0.6 nM). Exhibits selectivity for DGAT-1 over DGAT-2, Kv11.1 (hERG) and cytochrome P450 enzymes. Suppresses triacylglyceride (TAG) plasma excursion and adipose tissue TAG synthesis in rats. Reduces body weight of diet-induced obese rats. Cell permeable and orally bioavailable.
Specifications
Specifications
| Quantity | 10 mg |
| Product Type | AZD 3988 |
| Purity | >98% |
| Molecular Weight (g/mol) | 456.44 |
| Inhibitors | AZD 3988 |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction