missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZD 2858

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AZD 2858 is a potent glycogen synthase kinase-3 (GSK-3) inhibitor (IC50 values are 0.9 and 5 nM for GSK-3α and GSK-3β, respectively). AZD 2858 inhibits tau phosphorylation in vitro (IC50 = 76 nM), improves fracture healing and increases bone mass in rodent models of bone fracture. Also activates Wnt signaling in vitro and in vivo. Brain permeable and orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 3-Amino-6-[4-[(4-methyl-1-piperazinyl)sulfonyl]phenyl]-N-3-pyridinyl-2-pyrazinecarboxamide |
| CAS | 486424-20-8 |
| Quantity | 10 mg |
| Target | Glycogen Synthase Kinase Inhibitors |
| Molecular Formula | C21H23N7O3S |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction