missing translation for 'onlineSavingsMsg'
Learn More

AZD-2461, MedChemExpress

Product Code. 16491897
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16491897 50 mg 50mg
16486216 100 mg 100mg
16485738 10 mg 10mg
16408427 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491897 Supplier Medchem Express Supplier No. HY13536

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD-2461 is a potent PARP inhibitor, with IC50s of 5 nM, 2 nM and 200 nM for PARP1, PARP2 and PARP3, respectively.

AZD-2461 is a potent PARP inhibitor, with IC50s of 5 nM, 2 nM and 200 nM for PARP1, PARP2 and PARP3, respectively.

Specifications

Chemical Name or Material AZD-2461
CAS 1174043-16-3
Color White
Purity Grade Notes Research
Molecular Formula C22H22FN3O3
Quantity 50 mg
Solubility Information DMSO : ≥ 100 mg/mL (252.89 mM)
SMILES O=C1NN=C(CC2=CC=C(F)C(C(N3CCC(OC)CC3)=O)=C2)C4=C1C=CC=C4
Molecular Weight (g/mol) 395.43
Formula Weight 395.43
Percent Purity 97.3%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.