missing translation for 'onlineSavingsMsg'
Learn More

Azathioprine, TRC

Product Code. 30040964
Change view
Click to view available options
Quantity:
50 mg
500 mg
1 g
Unit Size:
1g
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040964 1 g 1g
30040722 500 mg 500mg
30040727 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040964 Supplier Toronto Research Chemicals Supplier No. TRCA8033501G
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-86-6
Molecular Formula C9 H7 N7 O2 S
Molecular Weight (g/mol) 277.26
Synonym Azathioprine, 6-[(1-Methyl-4-nitro-1H-imidazol-5-yl)sulfanyl]-7H-purine, 9H-Purine, 6-[(1-methyl-4-nitro-1H-imidazol-5-yl)thio]-, 1H-Purine, 6-[(1-methyl-4-nitro-1H-imidazol-5-yl)thio]-, Purine, 6-[(1-methyl-4-nitroimidazol-5-yl)thio]-, 6-(1-Methyl-4-nitroimidazol-5-yl)thiopurine, 6-(1-Methyl-4-nitromidazol-5-ylthio)purine, Azamune, Azanin, Azathioprin, Azathioprine, Azathropine, Azoran, Azothioprine, BW 57-322, Imuran, Imurek, Imurel, Muran, NSC 39084
IUPAC Name 6-(3-methyl-5-nitroimidazol-4-yl)sulfanyl-7H-purine
SMILES Cn1cnc(c1Sc2ncnc3nc[nH]c23)[N+](=O)[O-]

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H7N7O2S/c1-15-4-14-7(16(17)18)9(15)19-8-5-6(11-2-10-5)12-3-13-8/h2-4H,1H3,(H,10,11,12,13)
Formula Weight 277.0382
Percent Purity >95%
Chemical Name or Material Azathioprine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.