missing translation for 'onlineSavingsMsg'
Learn More

Azathioprine, MedChemExpress

Product Code. 16482109
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16482109 1 g 1g
16460848 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16482109 Supplier Medchem Express Supplier No. HYB02561g
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Azathioprine (BW 57-322) is an orally active immunosuppressive agent. Azathioprine can be converted in vivo to the active metabolite 6-mercaptopurine (6-MP). Azathioprine has myelosuppressive effects and induces apoptosis.

Azathioprine (BW 57-322) is an orally active immunosuppressive agent. Azathioprine can be converted in vivo to the active metabolite 6-mercaptopurine (6-MP). Azathioprine has myelosuppressive effects and induces apoptosis.

Specifications

Chemical Name or Material Azathioprine
CAS 446-86-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C9H7N7O2S
Quantity 1 g
Synonym BW 57-322
Solubility Information DMSO : 50 mg/mL (180.34 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.36 mM; Need ultrasonic)
SMILES CN1C(SC2=C3N=CNC3=NC=N2)=C([N+]([O-])=O)N=C1
Molecular Weight (g/mol) 277.26
Formula Weight 277.26
Percent Purity 99.26%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.