missing translation for 'onlineSavingsMsg'
Learn More

Azatadine dimaleate, MedChemExpress

Product Code. 16498378
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498378 10 mg 10mg
16498146 100 mg 100mg
16404277 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16498378 Supplier Medchem Express Supplier No. HYB0170A
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Azatadine dimaleate is an histamine and cholinergic inhibitor with IC50 of 6.5 nM and 10 nM, respectively.

Azatadine dimaleate is an histamine and cholinergic inhibitor with IC50 of 6.5 nM and 10 nM, respectively.

Specifications

Chemical Name or Material Azatadine dimaleate
CAS 3978-86-7
Color White
Purity Grade Notes Research
Molecular Formula C28H30N2O8
Quantity 10 mg
Synonym Azatadine maleate
Solubility Information H2O : ≥ 100 mg/mL (191.37 mM) ∣DMSO : ≥ 100 mg/mL (191.37 mM)
SMILES O=C(O)/C=C\C(O)=O.O=C(O)/C=C\C(O)=O.CN1CC/C(CC1)=C2C3=CC=CC=C3CCC4=CC=CN=C4\2
Molecular Weight (g/mol) 522.55
Formula Weight 522.55
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Mehr anzeigen Weniger anzeigen

Research purposes only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.