missing translation for 'onlineSavingsMsg'
Learn More

Azasetron hydrochloride, MedChemExpress

Product Code. 16464217
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464217 50 mg 50mg
16428338 10 mg 10mg
16428076 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16464217 Supplier Medchem Express Supplier No. HYB0068

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Azasetron (Y-25130) hydrochloride, a benzamide derivative, is a potent and selective 5-HT3 receptor antagonist. Azasetron is used in the study for Chemotherapy-induced nausea and vomiting (CINV).

Azasetron (Y-25130) hydrochloride, a benzamide derivative, is a potent and selective 5-HT3 receptor antagonist. Azasetron is used in the study for Chemotherapy-induced nausea and vomiting (CINV).

Specifications

Chemical Name or Material Azasetron hydrochloride
CAS 123040-16-4
Color White
Purity Grade Notes Research
Molecular Formula C17H21Cl2N3O3
Quantity 50 mg
Synonym Y-25130 hydrochloride
Solubility Information H2O : 20 mg/mL (51.78 mM; Need ultrasonic) ∣DMSO : 2.22 mg/mL (5.75 mM; Need ultrasonic)
SMILES O=C(C1=C(OCC(N2C)=O)C2=CC(Cl)=C1)NC3CN4CCC3CC4.[H]Cl
Molecular Weight (g/mol) 386.27
Formula Weight 386.27
Percent Purity 99.78%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.