missing translation for 'onlineSavingsMsg'
Learn More

Azaserine, MedChemExpress

Product Code. p-200096062
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16454507

Brand: Medchem Express HYB0919

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Azazerine (CI-337) is a competitive inhibitor of glutamine amidotransferase, a key enzyme responsible for glutamine metabolism.

Azazerine (CI-337) is a competitive inhibitor of glutamine amidotransferase, a key enzyme responsible for glutamine metabolism.

Specifications

Chemical Name or Material Azaserine
Physical Form Solid
CAS 115-02-6
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C5H7N3O4
For Use With (Application) Cancer-programmed cell death
Synonym CI-337 O-Diazoacetyl-L-serine P-165
Solubility Information H2O : 50 mg/mL (288.80 mM; Need ultrasonic)
SMILES N[C@@H](COC(C=[N+]=[N-])=O)C(O)=O
Molecular Weight (g/mol) 173.13
Formula Weight 173.13
Percent Purity 98.06%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.