missing translation for 'onlineSavingsMsg'
Learn More

Azaphen, MedChemExpress

Product Code. 16417996
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16417996

Brand: Medchem Express HYA0022

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress Pipofezine(Azafen or Azaphen) is a potent inhibitor of the reuptake of serotonin.

Pipofezine(Azafen or Azaphen) is a potent inhibitor of the reuptake of serotonin.

Specifications

Chemical Name or Material Azaphen
CAS 24853-80-3
Purity Grade Notes Research
Molecular Formula C16H21Cl2N5O
Quantity 100 mg
Synonym Azafen Pipofezin hydrochloride Pipofezine hydrochloride
SMILES [H]Cl.[H]Cl.CN1C2=CC=CC=C2OC3=C1C=C(N4CCN(C)CC4)N=N3
Molecular Weight (g/mol) 370.28
Formula Weight 370.28
Grade Research
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.