missing translation for 'onlineSavingsMsg'
Learn More

Azadiradione (~90%), TRC

Product Code. 30137193
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137193 10 mg 10mg
30134405 5 mg 5mg
30004453 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137193 Supplier Toronto Research Chemicals Supplier No. TRCA79734310MG
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26241-51-0
Molecular Formula C28 H34 O5
Molecular Weight (g/mol) 450.57
Synonym (5α,7α,13α,17α)-7-(Acetyloxy)-21,23-epoxy-4,4,8-trimethyl-24-norchola-1,14,20,22-tetraene-3,16-dione
IUPAC Name [(5R,7R,8R,9R,10R,13S,17R)-17-(3-furyl)-4,4,8,10,13-pentamethyl-3,16-dioxo-6,7,9,11,12,17-hexahydro-5H-cyclopenta[a]phenanthren-7-yl] acetate
SMILES CC(=O)O[C@@H]1C[C@H]2C(C)(C)C(=O)C=C[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](C(=O)C=C4[C@]13C)c5cocc5

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C28H34O5/c1-16(29)33-23-14-20-25(2,3)22(31)8-11-26(20,4)19-7-10-27(5)21(28(19,23)6)13-18(30)24(27)17-9-12-32-15-17/h8-9,11-13,15,19-20,23-24H,7,10,14H2,1-6H3/t19-,20+,23-,24-,26-,27-,28-/m1/s1
Formula Weight 450.2406
Chemical Name or Material Azadiradione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.