missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azadibenzocyclooctyne-PEG, MW 10,000, Thermo Scientific™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Azadibenzocyclooctyne-PEGis used for copper-free click chemistry reactions.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool,
Azadibenzocyclooctyne-PEGis used for copper-free click chemistry reactions.
Solubility
Soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool,
Specifications
Specifications
| Color | White to Gray |
| Quantity | 100 mg |
| Synonym | ADIBO-PEG, MW 10,000 |
| Solubility Information | Soluble in water. |
| Formula Weight | 10kDa |
| Physical Form | Solid |
| Chemical Name or Material | Azadibenzocyclooctyne-PEG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction