missing translation for 'onlineSavingsMsg'
Learn More

Azadibenzocyclooctyne-NHS ester, with 4 carbon linker

Product Code. 15430347
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15430347 100 mg 100mg
15420347 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15430347 Supplier Thermo Scientific Alfa Aesar Supplier No. J64128.MC
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Azadibenzocyclooctyne-NHS ester, with 4 carbon linker is used in various applications like protein-peptide conjugates, antibody-enzyme or antibody-drug conjugates, protein or peptide-oligonucleotide conjugates and surface modification. It is used to introduce cyclooctyne moiety in to biomolecules and amine containing compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azadibenzocyclooctyne-NHS ester, with 4 carbon linker is used in various applications like protein-peptide conjugates, antibody-enzyme or antibody-drug conjugates, protein or peptide-oligonucleotide conjugates and surface modification. It is used to introduce cyclooctyne moiety in to biomolecules and amine containing compounds.

Solubility
Soluble in dimethyl formamide, terahydrofuran, dimethyl sulfocide and dichloromethane.

Notes
Moisture and heat sensitive. Store in cool place. Incompatible with strong oxidizng agents.
TRUSTED_SUSTAINABILITY

Specifications

Color White to Gray
Quantity 100 mg
Sensitivity Moisture sensitive
Solubility Information Soluble in dimethyl formamide,terahydrofuran,dimethyl sulfocide and dichloromethane.
Formula Weight 402.4
Physical Form Solid
Chemical Name or Material Azadibenzocyclooctyne-NHS ester, with 4 carbon linker

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.