missing translation for 'onlineSavingsMsg'
Learn More

Azadibenzocyclooctyne-amine

Product Code. 15407009
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 15407009

Brand: Thermo Scientific Alfa Aesar J65637.MB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1255942-06-3 | C18H16N2O | 276.339 g/mol

Azadibenzocyclooctyne amine is a carbonyl reactive reagent used to incorporate ADIBO into organic compounds, surfaces or particlesAzadibenzocyclooctyne amine is widely useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. It reacts with azide functionalized compounds or bimolecules to give stable triazole linkage without a need for a Cu(I) catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azadibenzocyclooctyne amine is a carbonyl reactive reagent used to incorporate ADIBO into organic compounds, surfaces or particlesAzadibenzocyclooctyne amine is widely useful in strain-promoted copper-free azide-alkyne cycloaddition reactions. It reacts with azide functionalized compounds or bimolecules to give stable triazole linkage without a need for a Cu(I) catalyst.

Notes
Heat sensitive. Incompatible with amines and azides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1255942-06-3
Molecular Formula C18H16N2O
Molecular Weight (g/mol) 276.339
MDL Number MFCD22380759
InChI Key OCCYFTDHSHTFER-UHFFFAOYSA-N
Synonym dbco-amine, azadibenzocyclooctyne-amine, adibo-ch2 2-nh2.tfa, azadibenzocyclooctyne-ch2 2-amine.tfa, dibenzocyclooctyne-amine, for copper-free click chemistry, 5-beta-alanyl-11,12-didehydro-5,6-dihydrodibenzo b,f azocine, 3-amino-1-5-aza-3,4:7,8-dibenzocyclooct-1-yne-5-yl-1-propanone, 3-amino-1-2-azatricyclo 10.4.0.0?,? hexadeca-1 16 ,4,6,8,12,14-hexaen-10-yn-2-yl propan-1-one, 3-amino-1-2-azatricyclo 10.4.0.04,9 hexadeca-1 12 ,4 9 ,5,7,13,15-hexaen-10-yn-2-yl propan-1-one
PubChem CID 77078258
SMILES C1C2=CC=CC=C2C#CC3=CC=CC=C3N1C(=O)CCN

Specifications

Color Yellow to Orange
Quantity 25 mg
Formula Weight 276.33
Physical Form Solid
Chemical Name or Material Azadibenzocyclooctyne-amine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.