missing translation for 'onlineSavingsMsg'
Learn More

Azadibenzocyclooctyne acid

Product Code. 15496449
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 15496449

Brand: Thermo Scientific Alfa Aesar J64549.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1425485-72-8 | C21H19NO3 | 333.39 g/mol

Azadibenzocyclooctyne acid is the simplest amine reactive reagent for introducing a dibenzylcyclooctyne to available amine functionality.Azadibenzocyclooctyne acid is widely useful in strain-promoted copper-free azide-alkyne click chemistry reactions. It reacts with azide functionalized compounds or bimolecules to give stable triazole linkage without a need for a Cu(I) catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azadibenzocyclooctyne acid is the simplest amine reactive reagent for introducing a dibenzylcyclooctyne to available amine functionality.Azadibenzocyclooctyne acid is widely useful in strain-promoted copper-free azide-alkyne click chemistry reactions. It reacts with azide functionalized compounds or bimolecules to give stable triazole linkage without a need for a Cu(I) catalyst.

Solubility
Soluble in dimethyl sulfoxide and dimethyl formamide.

Notes
Store in cool place. Incompatible with amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1425485-72-8
Molecular Formula C21H19NO3
Molecular Weight (g/mol) 333.39
MDL Number MFCD22380737
InChI Key NIRLBCOFKPVQLM-UHFFFAOYSA-N
Synonym dbco-c6-acid, azadibenzocyclooctyne acid, dibenzocyclooctyne-acid, storage temp.:-20c, 6-11,12-didehydro-5,6-dihydrodibenzo b,f azocine-5-yl-6-oxohexanoic acid
PubChem CID 77078156

Specifications

Color White to Gray
Quantity 100 mg
Solubility Information Soluble in dimethyl sulfoxide and dimethyl formamide.
Formula Weight 333.38
Physical Form Solid
Chemical Name or Material Azadibenzocyclooctyne acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.