missing translation for 'onlineSavingsMsg'
Learn More

AZ876, MedChemExpress

Product Code. 16487488
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487488 10 mg 10mg
16441729 1 mg 1mg
16427456 100 mg 100mg
16423517 50 mg 50mg
16409977 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16487488 Supplier Medchem Express Supplier No. HY18282
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress AZ876 is a potent and high-affinity LXR agonist. AZ876 displays 25-fold and 2.5-fold more potent than GW3965 (HY-10627) on human (h)LXRα and hLXRβ respectively.

AZ876 is a potent and high-affinity LXR agonist. AZ876 displays 25-fold and 2.5-fold more potent than GW3965 (HY-10627) on human (h)LXRα and hLXRβ respectively.

Specifications

Chemical Name or Material AZ876
CAS 898800-26-5
Color Orange
Purity Grade Notes Research
Molecular Formula C24H29N3O3S
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (227.50 mM; Need ultrasonic)
SMILES O=C(C(NC1=CC=C(N2CCCCC2)C=C1)=C3C4=CC=CC=C4)N(C(C)(C)C)S3(=O)=O
Molecular Weight (g/mol) 439.57
Formula Weight 439.57
Percent Purity 99.26%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.