missing translation for 'onlineSavingsMsg'
Learn More

AZ7550 hydrochloride, MedChemExpress

Product Code. 16461068
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16461068

Brand: Medchem Express HYB0794A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

MedChemExpress AZ7550 hydrochloride is an active metabolite of AZD9291 and inhibits the activity of IGF1R with an IC50 of 1.6 μM.

AZ7550 hydrochloride is an active metabolite of AZD9291 and inhibits the activity of IGF1R with an IC50 of 1.6 μM.

Specifications

Chemical Name or Material AZ7550 hydrochloride
CAS 2309762-40-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H32ClN7O2
Quantity 5 mg
Solubility Information DMSO : 1.88 mg/mL (3.60 mM; Need ultrasonic and warming)
SMILES C=CC(NC1=CC(NC2=NC=CC(C3=CN(C)C4=C3C=CC=C4)=N2)=C(OC)C=C1N(C)CCNC)=O.[H]Cl
Molecular Weight (g/mol) 522.04
Formula Weight 522.04
Percent Purity 98.66%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.