missing translation for 'onlineSavingsMsg'
Learn More

AZ505, MedChemExpress

Product Code. 16498957
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498957 5 mg 5mg
16456676 100 mg 100mg
16452487 50 mg 50mg
16423466 25 mg 25mg
16406378 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16498957 Supplier Medchem Express Supplier No. HY15226
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AZ505 is a potent and selective SMYD2 inhibitor with an IC50 of 0.12 μM.

AZ505 is a potent and selective SMYD2 inhibitor with an IC50 of 0.12 μM.

Specifications

Chemical Name or Material AZ505
CAS 1035227-43-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H38Cl2N4O4
Quantity 5 mg
Solubility Information DMSO : ≥ 42 mg/mL (72.72 mM)
SMILES O=C(N(C1CCCCC1)CCNCCC2=C(OCC(N3)=O)C3=C(O)C=C2)CCNCCC4=CC=C(Cl)C(Cl)=C4
Molecular Weight (g/mol) 577.54
Formula Weight 577.54
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.