missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZ32, MedChemExpress
Description
AZ32 is an orally bioavailable and blood-brain barrier-penetrating ATM inhibitor with an IC50 of IC50 of 0.31 μM for ATM in cell.
Specifications
Specifications
| Chemical Name or Material | AZ32 |
| CAS | 2288709-96-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C20H16N4O |
| Quantity | 10 mg |
| Solubility Information | DMSO : ≥ 150 mg/mL (456.80 mM) |
| SMILES | O=C(NC)C(C=C1)=CC=C1C2=CN=C3C=NC(C4=CC=CC=C4)=CN32 |
| Molecular Weight (g/mol) | 328.37 |
| Formula Weight | 328.37 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?