missing translation for 'onlineSavingsMsg'
Learn More

AZ3146, MedChemExpress

Product Code. p-200098469
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496516 100 mg 100mg
16482267 50 mg 50mg
16406138 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16496516 Supplier Medchem Express Supplier No. HY14710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZ3146 is a reasonably potent and selective Mps1 inhibitor with IC50 of 35 nM for Mps1Cat.

AZ3146 is a reasonably potent and selective Mps1 inhibitor with IC50 of 35 nM for Mps1Cat.

Specifications

Chemical Name or Material AZ3146
CAS 1124329-14-1
Color Gray
Purity Grade Notes Research
Molecular Formula C24H32N6O3
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (220.97 mM; Need ultrasonic)
SMILES CN(CC1)CCC1OC2=CC=C(NC3=NC(N(C4CCCC4)C(N5C)=O)=C5C=N3)C(OC)=C2
Molecular Weight (g/mol) 452.55
Formula Weight 452.55
Percent Purity 97.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.