missing translation for 'onlineSavingsMsg'
Learn More

AZ191, MedChemExpress

Product Code. 16491287
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491287 50 mg 50mg
16457777 5 mg 5mg
16435796 100 mg 100mg
16415058 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16491287 Supplier Medchem Express Supplier No. HY12277
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AZ191 is a potent inhibitor that selectively inhibits DYRK1B with IC50 of 17 nM.

AZ191 is a potent inhibitor that selectively inhibits DYRK1B with IC50 of 17 nM.

Specifications

Chemical Name or Material AZ191
CAS 1594092-37-1
Color Brown
Purity Grade Notes Research
Molecular Formula C24H27N7O
Quantity 50 mg
Solubility Information DMSO : ≥ 30 mg/mL (69.85 mM)
SMILES CN1C2=CN=CC=C2C(C3=NC(NC4=CC=C(N5CCN(C)CC5)C=C4OC)=NC=C3)=C1
Molecular Weight (g/mol) 429.52
Formula Weight 429.52
Percent Purity 98.7%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.