missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZ Dyrk1B 33

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
AZ Dyrk1B 33 is a potent and selective ATP-competitive Dyrk1B kinase inhibitor (IC50 = 7 nM); displays distinct cellular effects when compared to DYRK1B knockdown through siRNA. Demonstrates cellular in vitro activity (IC50 = 194 nM). Exhibits better selectivity than AZ 191 (Cat. No 5232); displays no off-target effects against a panel of 124 kinases tested (no kinase was inhibited above 50% at 1 μM).
Specifications
Specifications
| Chemical Name or Material | 3-(2-Methyl-4-pyrimidinyl)-1-(phenylmethyl)-1H-pyrrolo[2,3-c]pyridine |
| CAS | 1679330-37-0 |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in 1eq. HCl and to 100mM in DMSO |
| Molecular Formula | C19H16N4 |
| Formula Weight | 300.36 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Purity | >98% |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction