missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZ 6102

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
AZ 6102 is a potent TNKS2 and TNKS1 inhibitor (IC50 values are 1 and 3 nM, respectively). Exhibits >100-fold selectivity for TNKS1/2 over PARP-1, PARP-2 and PARP-6. Inhibits Wnt signaling in DLD-1 cells (IC50 = 5 nM). Exhibits moderate oral bioavailability.
Specifications
Specifications
| Chemical Name or Material | rel-2-[4-[6-[(3R,5S)-3,5-Dimethyl-1-piperazinyl]-4-methyl-3-pyridinyl]phenyl]-3,7-dihydro-7-methyl-4H-pyrrolo[2,3-d]pyrimidin-4-one |
| CAS | 1645286-75-4 |
| Quantity | 10 mg |
| Target | Tankyrase Inhibitors |
| Molecular Formula | C25H28N6O |
| Formula Weight | 428.53 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction