missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AZ 6102
GREENER_CHOICE

Product Code. 18738208
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18738208

Brand: Tocris Bioscience™ 5775/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent TNKS1/2 inhibitor

AZ 6102 is a potent TNKS2 and TNKS1 inhibitor (IC50 values are 1 and 3 nM, respectively). Exhibits >100-fold selectivity for TNKS1/2 over PARP-1, PARP-2 and PARP-6. Inhibits Wnt signaling in DLD-1 cells (IC50 = 5 nM). Exhibits moderate oral bioavailability.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rel-2-[4-[6-[(3R,5S)-3,5-Dimethyl-1-piperazinyl]-4-methyl-3-pyridinyl]phenyl]-3,7-dihydro-7-methyl-4H-pyrrolo[2,3-d]pyrimidin-4-one
CAS 1645286-75-4
Quantity 10 mg
Target Tankyrase Inhibitors
Molecular Formula C25H28N6O
Formula Weight 428.53
Purity >98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.