missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZ 3146
Description
Potent and selective monopolar spindle 1 (Mps1) kinase inhibitor (IC50 = 35 nM). Displays selectivity over 46 other kinases including Cdk1 and aurora kinase B. Interferes with chromosome alignment and overrides spindle assembly checkpoint. Inhibits the recruitment of Mad1, Mad2 and centromere protein E (CENP-E) to kinetochores.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 452.55 |
| Purity | >99% |
| Molecular Weight (g/mol) | 452.559 |
| Chemical Name or Material | AZ 3146 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?