missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ AZ 12799734
GREENER_CHOICE

Product Code. p-7182920
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18735836

Brand: Tocris Bioscience™ 6870/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent TGF-βRI inhibitor

AZ 12799734 is a potent inhibitor of transforming growth factor-β type I receptor (TGF-βRI; ALK5) (IC50 = 17 nM in cellular inhibition assay and 47 nM in TGF-β-induced luciferase activity assay). Also inhibits ALK4, ALK6 and ALK7 at concentrations under 10 μM. Inhibits BMP signaling, phosphorylation of SMAD1 and SMAD2, and TGF-β-induced epithelial cell migration in vitro. Displays cardiotoxicity in vivo. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[[4-[(2.6-Dimethyl-3-pyridinyl)oxy]-2-pyridinyl]amino]benzenesulfonamide
CAS 1117684-36-2
Quantity 10 mg
Molecular Formula C18H18N4O3S
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.