missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AZ 12799734

Description
AZ 12799734 is a potent inhibitor of transforming growth factor-β type I receptor (TGF-βRI; ALK5) (IC50 = 17 nM in cellular inhibition assay and 47 nM in TGF-β-induced luciferase activity assay). Also inhibits ALK4, ALK6 and ALK7 at concentrations under 10 μM. Inhibits BMP signaling, phosphorylation of SMAD1 and SMAD2, and TGF-β-induced epithelial cell migration in vitro. Displays cardiotoxicity in vivo. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 4-[[4-[(2.6-Dimethyl-3-pyridinyl)oxy]-2-pyridinyl]amino]benzenesulfonamide |
| CAS | 1117684-36-2 |
| Quantity | 50 mg |
| Molecular Formula | C18H18N4O3S |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?