missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Tip Refill System, Empty Racks

Product Code. 11330285 Shop All Axygen Scientific Products
Change view
Click to view available options
Model:
RFL-R
RFL-R-1200
Packaging:
10 Racks/Pack; 3 Packs/Case
10 Racks/Pack; 5 Packs/Case
Unit Size:
Pack of 10
Pack of 30
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Packaging unitSize
11330285 RFL-R 10 Racks/Pack; 5 Packs/Case Pack of 10
12736719 RFL-R 10 Racks/Pack; 5 Packs/Case Pack of 50
11340285 RFL-R-1200 10 Racks/Pack; 3 Packs/Case Pack of 10
12746719 RFL-R-1200 10 Racks/Pack; 3 Packs/Case Pack of 30
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11330285 Supplier Axygen™ Supplier No. RFLR
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Easy-to-load refill racks for Axygen Tip Refill System. For use with P2/P10 Gilson-Style and 200μL Universal fit inserts.

Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.

  • RFL-R is suitable for both the 10 μL and 200 μL refill inserts
  • RFL-R-1200 is suitable for 1200 μL refill inserts
  • Certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Sterility Non-sterile
For Use With (Equipment) P2/P10 Gilson-Style and 200μL Universal fit inserts
Model RFL-R
Packaging 10 Racks/Pack; 5 Packs/Case
Product Type Empty Refill Rack with Lid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.