missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Tip Refill System

Product Code. 11320285 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
0.1 to 10 μL
1 to 200 μL
15 to 1250 μL
Unit Size:
Pack of 4608
Pack of 4800
Pack of 768
Pack of 960
This item is not returnable. View return policy

Product Code. 11320285

Brand: Axygen™ RFL300C

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Axygen™ style and universal fit pipet tips in easy-to-load refill racks

Axygen™ Tip Refill System contains durable refill rack that can be easily refilled by hand and is autoclavable. These translucent Racks includes color-coded inserts for tip identification and are certified RNase-/DNase free. They are available in different colors and are packaged in bulk or racks.
  • Certified to be RNase-/DNase-free
  • Nonpyrogenic
  • Translucent rack allows tips to be easily seen
  • Color-coded insert allows for easy identification of tip
  • TRUSTED_SUSTAINABILITY

    Specifications

    Volume (Metric) 0.1 to 10 μL
    Sterility Non-sterile
    Color Clear
    Autoclavable Autoclavable
    Packaging 96 Tips/Rack, 10 Racks/Pk, 5 Packs/Cs
    For Use With (Equipment) Gilson Pipetman P10, Gilson Pipetman P2, Universal Fit
    Graduated No
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.