missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Storage Microplates
Shop All Axygen Scientific Products
Click to view available options
Color:
Red
Unit Size:
Pack of 50
Description
Choose from a variety of configurations:
- 48 deep-well rectangular plate with pyramid bottom
- 96 deep-well plate with square wells
- 384 deep-well diamond plate, with square wells
- 96-well assay plate with round bottom (SBS footprint) or V-bottom (SBS footprint)
- 24 deep-well rectangular plate
Specifications
Specifications
| Certifications/Compliance | ANSI, SLAS 1-2004 |
| Format | Standard |
| Array | 8 x 12 |
| Autoclavable | Autoclavable |
| Color | Red |
| Material | Polypropylene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Well Shape | Conical Bottom (V-well) |
| For Use With (Application) | Sample collection, in-vitro growth chambers and long term storage |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction