missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Volume: 5 to 50 μL
- Clear, racked tips
- Sterile or Non-sterile
Order Info
- Microvolume tips:
- Racked: 384 tips/rack, 10 racks/pack, 5 packs/case
- Macrovolume tips:
- Bulk packaging: 250 tips/pack, 10 packs/case
Specifications
Specifications
| Volume (Metric) | 5 to 50 μL |
| Sterility | Non-sterile |
| Color | Clear |
| Autoclavable | Autoclavable |
| Clean Claims | DNase, RNase, ATP, and Endotoxin Free |
| Format | Rack |
| Tip Style | Microvolume |
| For Use With (Equipment) | Gilson Pipetman, Costar Onepette, Costar 8-Pette, Costar 12-Pette, Eppendorf, Thermo Scientific Finnpipette, Labpette Multichannel, Nichiryo, Oxford Benchmate, Rainin EDP2, Titertek |
| Material | Polypropylene |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?