missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Specialty Tips, for 16-Channel Pipette

Product Code. 11394255 Shop All Axygen Scientific Products
Change view
Click to view available options
Volume (Metric):
0.5 to 60 μL
Unit Size:
Pack of 3840
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11394255 0.5 to 60 μL Pack of 3840
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11394255 Supplier Axygen Supplier No. T384LR

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Suitable for Axypet-16 channel pipettor for 384/1536-well formats, Multi-channel Plus and macrovolume. With Maxymum Recovery.

  • Packaging: Rack
  • Color: Clear
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 0.5 to 60 μL
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, ATP, and Endotoxin Free
Format Rack
Tip Style Low Retention
For Use With (Equipment) Universal Fit
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.