Learn More
Axygen™ Septa Mat for ABI multi-capillary Sequencing Instrument
Product Code. 12587897 Shop All Axygen Scientific Products
Click to view available options
For Use With (Equipment):
ABI Multi-capillary Sequencing Instruments
Unit Size:
Pack of 50
Product Code. 12587897
Brand: Axygen™ AM96SEPTA3100
Septa Mat for ABI Multi-Capillary Sequencing Instrument
Axygen™ AxyMatsTM septa mat is designed to be used on ABI sequencer. Made of research-grade silicone, they can be autoclaved, and following a bleach wash and ethanol rinse protocol, they can also be reused.
  • Septa Mat for ABI Multi-Capillary Sequencing Instrument
  • For use with 96 PCR microplates
  • TRUSTED_SUSTAINABILITY
    Specifications
    For Use With (Equipment) ABI Multi-capillary Sequencing Instruments
    Product Type Septa Mat