missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Self-Standing Centrifuge Tubes

Product Code. 12729598 Shop All Axygen Scientific Products
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
Capacity (Metric):
2 mL
50 mL
Includes:
Amber screw cap
Screw cap
Unit Size:
Pack of 4000
Pack of 500
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Capacity (Metric) Includes unitSize
12729598 Sterile 50 mL Screw cap Pack of 500
12759588 Non-sterile 2 mL Amber screw cap Pack of 4000
11374195 Non-sterile 2 mL Amber screw cap Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 12729598 Supplier Axygen Supplier No. SCT50SS25S

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Graduated tubes with conical bottom

  • Writing area for sample identification
  • Caps close with one hand

TRUSTED_SUSTAINABILITY

Specifications

Cap Style Screw Cap
Autoclavable Not Autoclavable
Capacity (Metric) 50 mL
Color Clear
Material Polypropylene
For Use With (Application) For sample identification
Includes Screw cap
Packaging Bag
Sterility Sterile
Bottom Shape Conical
Max. RCF 12,000 x g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.