missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Writing area for sample identification
- Caps close with one hand
Specifications
Specifications
| Cap Style | Screw Cap |
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 2 mL |
| Color | Clear |
| Material | Polypropylene |
| For Use With (Application) | For sample identification |
| Includes | Amber screw cap |
| Sterility | Non-sterile |
| Bottom Shape | Conical |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?