missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Screw Caps without O-rings

Product Code. 11340315 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Green
Orange
Format:
Non-looped
Unit Size:
Pack of 4000
Pack of 500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Format unitSize
11340315 Orange Non-looped Pack of 500
12779348 Green Non-looped Pack of 4000
11300305 Green Non-looped Pack of 500
12719368 Orange Non-looped Pack of 4000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11340315 Supplier Axygen™ Supplier No. SCO

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Available in looped or non-looped format

  • Choose from a variety of colors

TRUSTED_SUSTAINABILITY

Specifications

Material Polypropylene
Color Orange
Format Non-looped
Sterility Non-sterile
For Use With (Equipment) Both skirted and non-skirted variations of the 0.5mL, 1.5mL, and 2mL microcentrifuge tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.