missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Screw Caps with O-rings

Product Code. 10387849 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Assorted
Blue
Brown
Clear
Gray
Green
Orange
Pink
Red
Violet
White
Yellow
Sterility:
Non-sterile
Sterile
Format:
Looped
Non-looped
Unit Size:
Pack of 4000
Pack of 500
48 product options available for selection
Product selection table with 48 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Sterility Format unitSize
10387849 Red Non-sterile Non-looped Pack of 4000
12759368 Brown Non-sterile Non-looped Pack of 4000
11306134 Brown Non-sterile Non-looped Pack of 500
11310325 Blue Non-sterile Non-looped Pack of 500
11300325 Assorted Non-sterile Non-looped Pack of 500
12508947 Brown Sterile Non-looped Pack of 500
12769368 Brown Sterile Non-looped Pack of 4000
12799368 Clear Sterile Non-looped Pack of 4000
12538947 Clear Sterile Non-looped Pack of 500
12518947 Blue Sterile Non-looped Pack of 500
15869072 Blue Sterile Non-looped -
11320325 Green Non-sterile Non-looped Pack of 500
10328609 Green Non-sterile Non-looped Pack of 4000
12789368 Clear Non-sterile Non-looped Pack of 4000
11366144 Clear Non-sterile Non-looped Pack of 500
12719378 Gray Non-sterile Non-looped Pack of 4000
11330325 Gray Non-sterile Non-looped Pack of 500
12729378 Gray Sterile Non-looped Pack of 4000
12769378 Brown Non-sterile Looped Pack of 4000
11360325 Brown Non-sterile Looped Pack of 500
12528957 Clear Sterile Looped Pack of 500
11300335 Orange Non-sterile Looped Pack of 500
12729388 Orange Non-sterile Looped Pack of 4000
12799378 Green Non-sterile Looped Pack of 4000
11350325 Blue Non-sterile Looped Pack of 500
12759378 Blue Non-sterile Looped Pack of 4000
12779378 Clear Non-sterile Looped Pack of 4000
12709388 Gray Non-sterile Looped Pack of 4000
12739388 Pink Non-sterile Looped Pack of 4000
11320335 Red Non-sterile Looped Pack of 500
12789388 Yellow Non-sterile Looped Pack of 4000
11350335 Yellow Non-sterile Looped Pack of 500
12709398 Orange Sterile Non-looped Pack of 4000
12799388 Orange Non-sterile Non-looped Pack of 4000
11360335 Orange Non-sterile Non-looped Pack of 500
12779388 White Non-sterile Looped Pack of 4000
12719398 Pink Non-sterile Non-looped Pack of 4000
11330335 Violet Non-sterile Looped Pack of 500
12729398 Pink Sterile Non-looped Pack of 4000
11390335 Violet Non-sterile Non-looped Pack of 500
12588967 Red Sterile Non-looped Pack of 500
11380335 Red Non-sterile Non-looped Pack of 500
12789398 White Sterile Non-looped Pack of 4000
12528977 White Sterile Non-looped Pack of 500
10692261 Yellow Non-sterile Non-looped Pack of 4000
11310345 Yellow Non-sterile Non-looped Pack of 500
12548977 Yellow Sterile Non-looped Pack of 500
12709408 Yellow Sterile Non-looped Pack of 4000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 48 options available.
48 options
This item is not returnable. View return policy
Product Code. 10387849 Supplier Axygen™ Supplier No. SCOR
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Select looped or non-looped format

Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.

  • Maximum G-force is 20,000 RCF.
  • RNase-/DNase-free
  • Nonpyrogenic


Tubes, with Cap, Sterile


All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.


Colored-cap versions are also available in sterile format.
TRUSTED_SUSTAINABILITY

Specifications

Includes O-ring
Material Polypropylene
Color Red
Format Non-looped
No. per Pack 500/Pk.
No. per Case 4000/Cs.
Sterility Non-sterile
For Use With (Equipment) Both skirted and non-skirted variations of the 0.5 mL, 1.5 mL, and 2 mL microcentrifuge tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.