Learn More
Axygen™ Screw Cap Tubes without Caps: Self-Standing with Conical Bottoms
Product Code. 11344225 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Clear
Sterility:
Sterile
Capacity (Metric):
1.5 mL
2 mL
Unit Size:
Pack of 4000
Pack of 500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Sterility Capacity (Metric) unitSize
11344225 Clear Sterile 1.5 mL Pack of 500
12739608 Clear Sterile 1.5 mL Pack of 4000
11394225 Clear Sterile 2 mL Pack of 500
12719618 Clear Sterile 2 mL Pack of 4000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11344225 Supplier Axygen™ Supplier No. ST150SSCS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Self-standing tubes
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Capacity (Metric) 1.5 mL
Color Clear
Material Polypropylene
Packaging Bag
Sterility Sterile
Closure Type Not Included
Bottom Shape Conical
Max. RCF 20,000 x g