Learn More
Axygen™ Screw Cap Tubes without Caps: Self-Standing with Conical Bottoms
Product Code. 11394225 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Clear
Sterility:
Sterile
Capacity (Metric):
1.5 mL
2 mL
Unit Size:
Pack of 4000
Pack of 500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Sterility Capacity (Metric) unitSize
11394225 Clear Sterile 2 mL Pack of 500
12739608 Clear Sterile 1.5 mL Pack of 4000
11344225 Clear Sterile 1.5 mL Pack of 500
12719618 Clear Sterile 2 mL Pack of 4000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11394225 Supplier Axygen™ Supplier No. ST200SSS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Self-standing tubes
Serve a broad range of laboratory needs including collection, processing, centrifugation, storage and packaging/shipping
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance RNase, DNase, pyrogen-free
Capacity (Metric) 2 mL
Color Clear
Material Polypropylene
Packaging Bag
Sterility Sterile
Closure Type Not Included
Bottom Shape Conical
Max. RCF 20,000 x g