Learn More
Axygen™ Screw Cap Tubes without Caps: Self-Standing
Product Code. 12789598 Shop All Axygen Scientific Products
Change view
Click to view available options
Color:
Amber
Clear
Sterility:
Non-sterile
Sterile
Capacity (Metric):
0.5 mL
1.5 mL
2 mL
Unit Size:
Pack of 4000
Pack of 500
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Sterility Capacity (Metric) unitSize
12789598 Amber Non-sterile 0.5 mL Pack of 4000
12759598 Clear Non-sterile 0.5 mL Pack of 4000
11384215 Clear Non-sterile 0.5 mL Pack of 500
12539117 Clear Sterile 0.5 mL Pack of 500
12779598 Clear Sterile 0.5 mL Pack of 4000
11394215 Amber Non-sterile 0.5 mL Pack of 500
11334225 Clear Non-sterile 1.5 mL Pack of 500
12729608 Clear Non-sterile 1.5 mL Pack of 4000
11354225 Amber Non-sterile 1.5 mL Pack of 500
12759608 Amber Non-sterile 1.5 mL Pack of 4000
12799608 Clear Non-sterile 2 mL Pack of 4000
11384225 Clear Non-sterile 2 mL Pack of 500
11304235 Amber Non-sterile 2 mL Pack of 500
12729618 Amber Non-sterile 2 mL Pack of 4000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Product Code. 12789598 Supplier Axygen™ Supplier No. ST050SSX
Only null left
Add to basket
Add to basket
Self-standing tubes
Axygen microcentrifuge tubes feature a screw cap that provides a tight secure seal. Screw cap tubes are available in a variety of colors with a conical or self-standing bottom. Sterile options are also available.
  • Maximum G-force is 20,000 RCF.
  • RNase-/DNase-free
  • Nonpyrogenic
Tubes, with Cap, Sterile
All sterile screw cap microcentrifuge tubes are pre-assembled with non-looped caps with O-Rings.
Colored-cap versions are also available in sterile format.
Order Info
Caps sold separately.
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Screw Cap
Certifications/Compliance RNase, DNase, pyrogen-free
Capacity (Metric) 0.5 mL
Color Amber
Material Polypropylene
No. per Pack 500/Pk.
Packaging 500 Tubes/Pk., 8 Packs/Cs.
Sterility Non-sterile
Graduated No
Closure Type Not Included
Bottom Shape Conical
Max. RCF 20,000 x g