missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Polypropylene Universal Fit Pipette Tip
Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
200 μL
Unit Size:
Pack of 4800
Description
- Polypropylene construction
- Nonpyrogenic
- RNase-/DNase-free
- Flexible walls
- Internal sealing rings to ensure a secure fit with less force required to load and eject
- Bevelled
Order Info
dimProductType
Specifications
Specifications
| Volume (Metric) | 200 μL |
| Color | Yellow |
| Clean Claims | DNase, RNase, and Pyrogen Free |
| For Use With (Application) | with almost all brands of single and multichannel pipettes |
| Format | Rack |
| Tip Style | Universal |
| For Use With (Equipment) | Universal Fit |
| Material | Polypropylene |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction